Water Pollution Standard, Turbidity, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
777 893
CA11022-896EA 357.16 CAD
CA11022-896 CA11027-934
Water Pollution Standard, Turbidity, ERA (Waters)
Standards Environmental Analysis Standards
One 15 mL screw-cap vial yields up to 1 L of this water pollution, turbidity standard after dilution
Use with nephelometric methods.