Soil Standard, Metals in Soil, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
540 620
CA11020-080EA 1405.46 CAD
CA11020-080 CA11027-924
Soil Standard, Metals in Soil, ERA (Waters)
Standards Environmental Analysis Standards
One 40 g metals in soil, soil sample in a screw-cap bottle can be used for all ICP, AA, RCRA and Superfund methods, including EPA digestion methods 3050 hotplate and 3051 microwave
This standard includes all of the metals listed below.