Water Supply Standard, Hardness, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
555 693
CA11020-054EA 363.98 CAD
CA11020-054 CA11020-056
Water Supply Standard, Hardness, ERA (Waters)
Standards Environmental Analysis Standards
This whole-volume standard is ready-to-use as provided
It requires no dilution before analysis. One 250 mL whole-volume bottle of this water supply, hardness standard is ready to analyze.