Water Supply Standard, Mercury, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
666 551
CA11020-074EA 248.3 CAD
CA11020-074 CA11022-734
Water Supply Standard, Mercury, ERA (Waters)
Standards Environmental Analysis Standards
One 15 mL screw-cap vial yields up to 1 L of this water supply, mercury standard after dilution
Use with CVAA, ICP-MS or CVAFS methods.