Water Supply Standard, Haloacetic Acids, ERA
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
852 684
CA11020-052EA 503.4 CAD
CA11020-052 CA11022-698
Water Supply Standard, Haloacetic Acids, ERA
Standards Environmental Analysis Standards
One 2 mL flame-sealed ampule yields up to 2 L of this water supply, haloacetic acids (HAA) standard after dilution
Bromochloroacetic Acid, Dibromoacetic Acid, Dichloroacetic Acid, Monobromoacetic Acid, Monochloroacetic Acid, Trichloroacetic Acid
Use with EPA method 552. This standard includes all of the analytes listed below at 5 to 50 µg/L.