Air and Emission Standard, Mercury in Impinger Solution, ERA
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
1128 1028
CA11020-072EA 544.33 CAD
CA11020-072 CA11022-738
Air and Emission Standard, Mercury in Impinger Solution, ERA
Standards Environmental Analysis Standards
One 15 mL screw-cap vial contains approximately 14 mL mercury in impinger solution, standard concentrate for use with EPA methods 20 and 101A.