Water Pollution Standard, Diesel Range Organics, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
641 764
CA11020-048EA 598.68 CAD
CA11020-048 CA11020-050
Water Pollution Standard, Diesel Range Organics, ERA (Waters)
Standards Environmental Analysis Standards
One 2 mL flame-sealed ampule yields up to 2 L of this water pollution, diesel range organics (DRO) standard in water after dilution
Use with modified EPA 8015 GC/FID methods. This standard includes #2 diesel at 800 to 6,000 µg/L.
Order Now
SPECIFICATIONS
Environmentally Preferable
Concentration 800-6,000 µg/L