Water Supply Standard, Cyanide, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
983
556
CA11020-042EA
333.41
CAD
CA11020-042
CA11022-660
Water Supply Standard, Cyanide, ERA (Waters)
Standards
Environmental Analysis Standards
One 15 mL screw-cap vial yields up to 2 L of this water supply, cyanide standard after dilution
The source material of this standard is free cyanide.