Water Pollution Standard, Chlorinated Acid Herbicides, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
829 718
CA11020-028EA 472.85 CAD
CA11020-028 CA11022-650
Water Pollution Standard, Chlorinated Acid Herbicides, ERA (Waters)
Standards Environmental Analysis Standards
One 2 mL flame-sealed ampule yields up to 2 L of this water pollution, chlorinated acid herbicides standard after dilution
Acifluorfen, Bentazone, Chloramben, 2,4-D, 2,4-DB, Dacthal diacid (DCPA), Dalapon, Dicamba, 3,5-Dichlorobenzoic acid, Dichlorprop, Dinoseb, MCPA, MCPP, 4-Nitrophenol, Pentachlorophenol, Picloram, 2,4,5-T, 2,4,5-T P (Silvex).
Use with EPA methods 615 and 8151. This standard contains a subset of analytes listed below at 2 to 10 µg/L. The analytes MCPA and MCPP are listed at 10 to 100 µg/L.
Ordering information: 4-nitrophenol and pentachlorophenol are not within the EPA/NELAC range. Use with the water pollution, acids standard, ERA Cat. No 712_C.