Water Pollution Standard, Bromide, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
769 887
CA11020-026EA 333.41 CAD
CA11020-026 CA11027-928
Water Pollution Standard, Bromide, ERA (Waters)
Standards Environmental Analysis Standards
One mL screw-cap vial yields up to 2 L of this water pollution, bromide standard after dilution
Use with ion chromatography or colorimetric methods.