Hydrometer Racks, Polypropylene, Dynalon
Supplier: Dynalon
103215-0001 103215-0000
30620-398EA 247.58 CAD
30620-398 30620-396
Hydrometer Racks, Polypropylene, Dynalon
Racks Hydrometer Racks
The 18-place polypropylene racks are specially designed to safeguard glass hydrometers.
  • The three tiered rack with handles allows easy transport and storage of hydrometers
  • Top two plates have 1" diameter holes while the bottom plate has wells which taper from 1" to ⅜"
Tall 7" and short 5⅜" version available
Ordering information: Additional sizes and materials are available upon request.