VWR® Metzenbaum Dissecting Scissors
Supplier: VWR
11648-343
89259-982EA 153.32 CAD
89259-982
VWR® Metzenbaum Dissecting Scissors
Scissors
VWR® Metzenbaum Dissecting Scissors are very delicate stainless steel scissors with curved blades.
  • Stainless steel fabrication
Metzenbaum scissors are surgical scissors designed for cutting delicate tissue and blunt dissection. Scissors come in variable lengths and have a relatively long shank-to-blade ratio.