Water Pollution Standard, Base/Neutrals, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
833 711
CA11020-024EA 625.84 CAD
CA11020-024 CA11022-634
Water Pollution Standard, Base/Neutrals, ERA (Waters)
Standards Environmental Analysis Standards
One 2 mL flame-sealed ampule yields up to 2 L of this water pollution, bases and neutrals standard after dilution
Acenaphthene, Acenaphthylene, 2-Amino-1-methylbenzene, (o-Toluidine), Aniline, Anthracene, Benzidine, Benzo(a)anthracene, Benzo(b)fluoranthene, Benzo(k)fluoranthene, Benzo(g,h,i)perylene, Benzo(a)pyrene, Benzyl alcohol, 4-Bromophenyl phenyl ether, Butyl benzyl phthalate, Carbazole, 4-Chloroaniline, bis(2-Chloroethoxy)methane, bis(2-Chloroethyl)ether, bis(2-Chloroisopropyl)ether, 1-Chloronaphthalene, 2-Chloronaphthalene, 4-Chlorophenyl phenyl ether, Chrysene, Dibenz(a,h)anthracene, Dibenzofuran, 1,2-Dichlorobenzene, 1,3-Dichlorobenzene, 1,4-Dichlorobenzene, 3,3’-Dichlorobenzidine, Diethyl phthalate, Dimethyl phthalate, Di-n-butyl phthalate, 2,4-Dinitrotoluene, 2,6-Dinitrotoluene, Di-n-octyl phthalate, bis(2-Ethylhexyl)phthalate, Fluoranthene, Fluorene, Hexachlorobenzene, Hexachlorobutadiene, Hexachlorocyclopentadiene, Hexachloroethane, Indeno(1,2,3-cd)pyrene, Isophorone, 2-Methylnaphthalene, Naphthalene, 2-Nitroaniline, 3-Nitroaniline, 4-Nitroaniline, Nitrobenzene, N-Nitrosodiethylamine, N-Nitrosodimethylamine, N-Nitroso-di-n-propylamine, N-Nitrosodiphenylamine, Pentachlorobenzene, Phenanthrene, Pyrene, Pyridine, 1,2,4,5-Tetrachlorobenzene, 1,2,4-Trichlorobenzene
Use with EPA methods 625 and 8270. This standard contains a subset of the analytes listed below at 10 to 225 µg/L. The analyte Benzidine is listed at 100 to 1,000 µg/L.