VWR® Aluminum Test Tube Racks
Supplier: VWR International
89259-894 89259-896 89259-898
89259-894EA 62.09 CAD
89259-894 89259-896 89259-898
VWR® Aluminum Test Tube Racks
Racks Tube Racks
VWR® Aluminum Test Tube Racks are punched from hard sheet aluminum.
  • Edges and holes have a smooth finish
  • Practical, rust-proof design
S-shaped rack is practical and rust-proof. The low profile and wide, flat base prevent rack from tipping. Rack also slides into other racks for compact storage.