Soil Standard, TCLP Metals in Soil, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
544 629
CA11020-128EA 1558.5 CAD
CA11020-128 CA11022-874
Soil Standard, TCLP Metals in Soil, ERA (Waters)
Standards Environmental Analysis Standards
One 105 g TCLP metals in soil, soil standard in a screw-cap bottle is designed specifically to meet all state requirements for TCLP extraction and analysis for the metals listed below.
Antimony, Arsenic, Barium, Beryllium, Cadmium, Chromium, Lead, Mercury, Nickel, Selenium, Silver, Zinc