VWR®, Aluminum Diamond Mesh Test Tube Baskets, Rectangular
Supplier: VWR International
89259-774 89259-776 89259-778 89259-780 89259-782 89259-784 89259-834 89259-836 89259-960
89259-774EA 56.98 CAD
89259-774 89259-776 89259-778 89259-780 89259-782 89259-784 89259-834 89259-836 89259-960
VWR®, Aluminum Diamond Mesh Test Tube Baskets, Rectangular
Baskets Baskets for Test Tubes
VWR® Aluminum Diamond Mesh Baskets are made of heavy-gauge expanded aluminum that has been riveted for a longer life.
  • Rigid construction
  • Wire mesh allows full flow-thrugh of cleaning solution
  • Safe handling with no sharp edges
Strip bands cover sharp edges all around for safe handling. Expanded metal construction provides rigidity and full flow-thru of the cleaning solution.