Soil Standard, Cyanide in Soil, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
541 621
CA11022-662EA 748.68 CAD
CA11022-662 CA11022-664
Soil Standard, Cyanide in Soil, ERA (Waters)
Standards Environmental Analysis Standards
One 40 g cyanide in soil, soil standard in a screw-cap bottle is ready for use in all distillation and colorimetric methods.