Calibration Standard, Phenol, ERA
Supplier: ERA (Waters)
982
CA11022-832EA 496.8 CAD
CA11022-832
Calibration Standard, Phenol, ERA
Standards Environmental Analysis Standards
Calibration standards can be used for primary calibration or to prepare second source calibration check standards
One 500 mL amber glass bottle of this phenol calibration standard is preserved with H2SO4 at 1,000 mg/L.
They are traceable to NIST standard reference materials, where available, and are guaranteed stable for one year.
Ordering information: Certification documentation includes manufacturing uncertainties, traceability summaries and densities to aid in performing quantitative dilutions. The documentation for metal standards includes impurities.