Water Pollution Standard, Demand, 500 ml, ERA (Waters)
Supplier: ERA (Waters)
743
CA11022-668EA 466.21 CAD
CA11022-668
Water Pollution Standard, Demand, 500 ml, ERA (Waters)
Standards Environmental Analysis Standards
This whole-volume standard is ready-to-use as provided
It requires no dilution before analysis. One 500 mL whole-volume bottle of this water pollution, demand standard is ready to analyze.
Ordering information: This demand standard is guaranteed stable for one month after receipt at your facility.