Water Pollution Standard, Trace Metals, 500 ml, ERA (Waters)
Supplier: ERA (Waters)
740
CA11022-892EA 561.48 CAD
CA11022-892
Water Pollution Standard, Trace Metals, 500 ml, ERA (Waters)
Standards Environmental Analysis Standards
This whole-volume standard is ready-to-use as provided
It requires no dilution before analysis. One 500 mL whole-volume bottle of this water pollution, trace metals standard is ready to analyze. Use with AA, ICP-OES or ICP-MS methods.
Ordering information: This trace metals standard is guaranteed stable for one month after receipt at your facility.