Water Pollution Standard, Low-Level Total Residual Chlorine, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
881 917
CA11022-730EA 357.16 CAD
CA11022-730 CA11022-732
Water Pollution Standard, Low-Level Total Residual Chlorine, ERA (Waters)
Standards Environmental Analysis Standards
This low-level total residual chlorine standard is designed for testing at low µg/L levels
One 2 mL flame-sealed ampule yields up to 2 L after dilution. Use with sensitive titrimetric or colorimetric methods.