Septa, Thermo Scientific
Supplier: Thermo Scientific
313P3240 313P3233 313P3211 313B3240 313B3233 31303215-BP 31303250-BP 31303230-BP 31303233-BP 31303240-BP 313G3215-BP 313G3230-BP 313G3240-BP 313P3215-BP 313P3233-BP 313P3240-BP
10056-064EA 59.46 CAD
10056-064 10056-066 10056-068 10056-070 10056-080 10056-082 10056-084 10056-086 10056-088 10056-090 10057-432 10057-434 10057-438 10057-440 10057-442 10141-480 10141-482 10141-484 10141-486 10141-488 10141-490 10141-494 10141-496 10141-498 10141-502 10141-504
Septa, Thermo Scientific
Septa
Thermo Scientific™ BTO and TR-Green septa for GC instruments.
  • Variety of materials and temperature limits for all applications
  • For use in Thermo Scientific™ TRACE™ and FOCUS™ GCs and instruments from other manufacturers
  • Contaminant-free in individual blister packs
Made of low-bleed silicone, these septa have excellent mechanical properties, are ideal for demanding GC-MS applications, and may be used reliably up to 400 °C.
Order Now
SPECIFICATIONS
Environmentally Preferable