Water Pollution Standard, Total Residual Chlorine, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
501 587
CA11020-138EA 272.08 CAD
CA11020-138 CA11022-890
Water Pollution Standard, Total Residual Chlorine, ERA (Waters)
Standards Environmental Analysis Standards
One 2 mL flame-sealed ampule yields up to 2 L of this water pollution, total residual chlorine standard after dilution
Use with titrimetric or colorimetric methods.