Water Pollution Standard, Surfactants-MBAS, ERA
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
892 776
CA11020-126EA 452.43 CAD
CA11020-126 CA11022-872
Water Pollution Standard, Surfactants-MBAS, ERA
Standards Environmental Analysis Standards
One 15 mL screw-cap vial yields up to 2 L of this water pollution, surfectants (MBAS) standard after dilution
Use for the analysis of surfectants-MBAS with EPA method 425.1.