Water Pollution Standard, Sulfide, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
891 71
CA11020-124EA 466.01 CAD
CA11020-124 CA11022-870
Water Pollution Standard, Sulfide, ERA (Waters)
Standards Environmental Analysis Standards
One 10 mL flame-sealed ampule yields up to 1 L of this water pollution, physical property standard after dilution
Use for the analysis of sulfide by titrimetric or colorimetric methods or ISE.
Ordering information: Preserved sample is guaranteed stable.