Water Pollution Standard, Silica, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
775 890
CA11022-850EA 326.59 CAD
CA11022-850 CA11027-932
Water Pollution Standard, Silica, ERA (Waters)
Standards Environmental Analysis Standards
One 60 mL poly bottle yields up to 1 L of this water pollution, silica standard after dilution
Use for the analysis of silica (SiO2) with colorimetric or ICP methods.