MAbPac™ SEC-1 Columns, Thermo Scientific
Supplier: Thermo Scientific
074696 074697 075592 088790 088460 088789 088460
10043-316EA 4606.79 CAD
10043-316 10043-318 10043-334 10148-482 10148-562 10148-564 10159-108 10852-498
MAbPac™ SEC-1 Columns, Thermo Scientific
Chromatography Columns
The proprietary hydrophilic bonded layer in these MAbPac SEC-1 Columns results in minimal non-desired interactions between the proteins and the stationary phase.
The Thermo Scientific™ MAbPac™ SEC-1 is a size exclusion chromatography (SEC) column designed for mAb characterization, including the separation of monomers, aggregates, and fragments. The unique chemistry is stable under both denaturing and non-denaturing conditions, using high-salt, low-salt, or volatile mobile phases.
The MAbPac SEC-1 column is based on high-purity, spherical, porous (300 Å), 5 μm silica particles covalently modified with a proprietary diol hydrophilic layer. The column is packed into three different int.Ø formats (7.8, 4.0, and 2.1 mm) to accommodate different applications and sample loadings. The 7.8 mm int.Ø column provides the highest resolution of mAb and aggregates separation and rugged method for routine analysis. The 4.0 mm int.Ø column offers excellent resolution at greatly reduced mobile phase consumption and requires modern HPLC system with minimal extra column volumes. The 2.1 mm int.Ø column provides the highest sensitivity for small amount of sample loading, and is suitable for direct MS detection.
Columns feature rugged, reproducible packing. Stable surface bonding leads to low column bleed and compatibility with MS, ELSD and Thermo Scientific™ Dionex™ Corona™ Aerosol Detection. These columns provide superior performance for the analysis of monoclonal antibodies, aggregates, and their fragments.