Water Pollution Standard, Tin and Titanium, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
517
573
CA11020-130EA
289.23
CAD
CA11020-130
CA11027-920
Water Pollution Standard, Tin and Titanium, ERA (Waters)
Standards
Environmental Analysis Standards
One 15 mL screw-cap vial yields up to 1 L of this water pollution, tin and titanium standard after dilution
Use with AA, ICP-OES or ICP-MS methods.