Water Pollution Standard, Trace Metals, 15 ml, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
500 586
CA11020-140EA 391.33 CAD
CA11020-140 CA11027-922
Water Pollution Standard, Trace Metals, 15 ml, ERA (Waters)
Standards Environmental Analysis Standards
One 15 mL screw-cap vial yields up to 1 L of this water pollution, trace metals standard after dilution
Use with AA, ICP-OES or ICP-MS and select colorimetric methods.