Air and Emission Standard, Metals on Filter Paper, ERA
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
1025 1125
CA11020-082EA 748.49 CAD
CA11020-082 CA11022-746
Air and Emission Standard, Metals on Filter Paper, ERA
Standards Environmental Analysis Standards
One filter paper sample, packaged in a 50 mm polystyrene petri dish contains a single 47 mm tissue quartz filter
This metals on filter paper standard is ready for use with EPA method 29.