Radiochemistry Standard, Iodine-131, ERA (Waters)

Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)

810 750
CA11022-726EA 2565.21 CAD
CA11022-726 CA10344-928
Radiochemistry Standard, Iodine-131, ERA (Waters)
Standards Environmental Analysis Standards

One 12 mL screw-top vial yields up to 2 liters after dilution.


Contains Iodine-131 within the range 3-30 pCi/L.
Due to short half-life, CRMs, PTs and QRs are available only during January, April, July and October.

Order Now

SPECIFICATIONS

Environmentally Preferable
Method EPA 901.1, EPA 902.0
Shelf life min. 12 months (on ship date)
Storage temperature 20 - 25°C (Ambient)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR