Radiochemistry Standard, Strontium-89/90, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
807 757
CA11022-862EA 2330.45 CAD
CA11022-862 CA10344-932
Radiochemistry Standard, Strontium-89/90, ERA (Waters)
Standards Environmental Analysis Standards
One 12 mL screw-top vial yields up to 2 liters after dilution.
Analytes:
Strontium-89 10 - 70 pCi/L
Strontium-90 3 - 45 pCi/L