Radiochemistry Standard, Water Tritium, MRAD, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
806 616 616QR
CA11022-916EA 1425.54 CAD
CA11022-916 CA10344-600 CA10345-564
Radiochemistry Standard, Water Tritium, MRAD, ERA (Waters)
Standards Environmental Analysis Standards
Sample set includes two 125 mL whole volume bottles ready to analyze as received. One Sample for Tritium analysis and one for blank correction.
Analyte
Tritium: 3000 - 30000 pCi/L