Radiochemistry Standard, Water Gross Alpha/Beta, MRAD, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
805 615QR
CA11022-912EA 1592.15 CAD
CA11022-912 CA10345-562
Radiochemistry Standard, Water Gross Alpha/Beta, MRAD, ERA (Waters)
Standards Environmental Analysis Standards
One 12 mL screw-top vial yields up to 2 liters after dilution. Includes the radionuclides below.
Analytes
Gross Alpha: 10 - 200 pCi/L
Gross Beta: 10 - 200 pCi/L