Radiochemistry Standard, Air Filter Gross Alpha/Beta MRAD, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
801 607 607QR
CA11022-620EA 1450.25 CAD
CA11022-620 CA10344-596 CA10345-556
Radiochemistry Standard, Air Filter Gross Alpha/Beta MRAD, ERA (Waters)
Standards Environmental Analysis Standards
Acrylic treated 47 mm diameter glass fiber filter.
Analyte: Gross Alpha 5-100 pCi/Filter