Radiochemistry Standard, Air Filter Radionuclides, MRAD, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
One 47 mm diameter glass fiber filter contains the alpha, beta and gamma emitting radionuclides listed below.
Analytes
Americium-241: 2 - 80 pCi/Filter
Cesium-134: 50 - 1500 pCi/Filter
Cesium-137: 50 - 1500 pCi/Filter
Cobalt-60: 50 - 1500 pCi/Filter
Iron-55: 50 - 1500 pCi/Filter
Manganese-54: 50 - 1500 pCi/Filter
Plutonium-238: 2 - 80 pCi/Filter
Plutonium-239: 2 - 80 pCi/Filter
Strontium-90: 5 - 200 pCi/Filter
Uranium (mass): 6 - 240 µg/Filter
Uranium-234: 2 - 80 pCi/Filter
Uranium-238: 2 - 80 pCi/Filter
Uranium-Total: 4 - 160 pCi/Filter
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...