Radiochemistry Standard, Gross Alpha/Beta, RAD, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
809 759 759QR
CA11022-692EA 3085.79 CAD
CA11022-692 CA11022-694 CA10345-652
Radiochemistry Standard, Gross Alpha/Beta, RAD, ERA (Waters)
Standards Environmental Analysis Standards
This radiochemistry standard is provided as a convenient, easy-to-prepare concentrate
One 12 mL screw-cap vial yields up to 1 L of this radiochemistry, gross alpha/beta standard after dilution.
Analytes: Gross Alpha 7-75 pCi/L