Radiochemistry Standard, Gamma Emitters, RAD, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
808
758
758QR
CA11022-686EA
2330.45
CAD
CA11022-686
CA11022-688
CA10345-650
Radiochemistry Standard, Gamma Emitters, RAD, ERA (Waters)
Standards
Environmental Analysis Standards
This radiochemistry standard is provided as a convenient, easy-to-prepare concentrate
One 12 mL screw-cap vial yields up to 2 L of this radiochemistry, gamma emitters standard after dilution.
Analytes
Barium-133: 10 - 100 pCi/L
Cesium-134: 10 - 100 pCi/L
Cesium-137: 20 - 240 pCi/L
Cobalt-60: 10 - 120 pCi/L
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...