Radiochemistry Standard, Naturals, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
811
751
751QR
CA11022-750EA
2565.21
CAD
CA11022-750
CA11022-752
CA10345-646
Radiochemistry Standard, Naturals, ERA (Waters)
Standards
Environmental Analysis Standards
This radiochemistry standard is provided as a convenient, easy-to-prepare concentrate
One 12 mL screw-cap vial yields up to 8 L of this radiochemistry, naturals standard after dilution.
Analytes
Radium-226: 1 - 20 pCi/L
Radium-228: 2 - 20 pCi/L
Uranium (activity): 2 - 70 pCi/L
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...