E-A-R™ Classic™ SuperFit™ Earplugs, 3M™
Supplier: Avantor
CA89006-772CS 647.26 CAD
CA89006-772 CA89006-804 CA89006-820 CA89006-822 CA89006-840
E-A-R™ Classic™ SuperFit™ Earplugs, 3M™
Earplugs Disposable Earplugs
3M™ E-A-R Classic™ SuperFit™ Earplugs feature an orange fitting ring that acts as a built-in visual check.
If the ring is visible after insertion, refitting is recommended for best hearing protection.
Order Now
SPECIFICATIONS
Environmentally Preferable