Water Pollution Standard, QC Plus – Fluoride, ERA (Waters)
Supplier: ERA (Waters)
4423
CA11022-682EA 241.53 CAD
CA11022-682
Water Pollution Standard, QC Plus – Fluoride, ERA (Waters)
Standards Environmental Analysis Standards
ERA's QC Plus program includes environmental analytes at concentrations that reflect realistic levels of pollutants in industrial settings
One 15 mL screw-cap vial yields up to 2 L of this water pollution, QC Plus Fluoride standard after dilution.
Each sample level is designed for wastewater and industrial analysis. These QC samples are an asset to any quality assurance program because they enable you to test your internal systems to ensure that your equipment, methods and analysts are producing quality data.