Water Supply Standard, Halomethanes (THMs), ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
702 842
CA11022-700EA 445.67 CAD
CA11022-700 CA11022-702
Water Supply Standard, Halomethanes (THMs), ERA (Waters)
Standards Environmental Analysis Standards
One 2 mL flame-sealed ampule of this water supply, halomethanes (THMs) standard yields in excess of 200 mL after dilution
Bromodichloromethane, Bromoform, Chlorodibromomethane, Chloroform
Use with EPA methods 502.2, 524.2 and 551. This standard contains all of the analytes listed below at 5 to 50 µg/L.