Water Supply Standard, Residual Chlorine, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
593 696
CA11020-108EA 302.62 CAD
CA11020-108 CA11020-110
Water Supply Standard, Residual Chlorine, ERA (Waters)
Standards Environmental Analysis Standards
One 2 mL flame-sealed ampule yields up to 2 L of this water supply, residual chlorine standard after dilution.