Combination Lockout Toolbox, Brady Worldwide®
Supplier: Avantor
CA89008-128KT 396.91 CAD
CA89008-128 CA89008-130
Combination Lockout Toolbox, Brady Worldwide®
Security Locks
The combination lockout toolbox gives users all the basics for locking out gate and ball valves, breakers, switches, and plugs.
Kit components include: one Mini Cable Lockout, one 120 V Snap-On Breaker Lockout, one 120/277 V Clamp-On Breaker Lockout, one 480/600 V Clamp-On Breaker Lockout, one Universal Multi-Pole Breaker Lockout, one Cleat for 120/277 V Clamp-On Lockout, one Cleat for 480/600 V Clamp-On Lockout, one 110 V Plug Lockout, one Wall Switch Lockout, one 2.54 to 16.51 cm (1 to 61/2") Adjustable Gate Valve Lockout, one 6.35 to 25.4 mm (1/4 to1") Ball Valve Lockout, one 3.175 to 7.62 cm (11/4 to 3") Ball Valve Lockout, one Lockout Belt Pouch, and one Extra-Large Lockout Toolbox.