Water Pollution Standard, Organophosphorus Pesticides (OPP), ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
934 665
CA11020-100EA 598.68 CAD
CA11020-100 CA11022-794
Water Pollution Standard, Organophosphorus Pesticides (OPP), ERA (Waters)
Standards Environmental Analysis Standards
One 2 mL flame-sealed ampule yields up to 2 L of this water pollution, organophosphorus pesticides (OPP) standard after dilution
Azinphos-methyl (Guthion), Carbophenothion, Chlorpyrifos, Demeton O & S, Diazinon, Dichlorvos (DDVP), Dimethoate, Dioxathion, Disulfoton, Ethion, Ethoprop, Ethyl Parathion (Parathion), Famphur, Fonofos, Malathion, Methyl parathion, Phorate, Phosmet, Ronnel, Stirophos (tetrachlorovinphos), Terbufos.
Use with EPA methods 614, 622 and 8141. This standard contains a subset of the analytes listed below at 2 to 20 µg/L.