Water Pollution Standard, Nitrogen Pesticides, ERA (Waters)
Supplier: ERA (Waters)
487 674
CA11020-094EA 625.84 CAD
CA11020-094 CA11022-768
Water Pollution Standard, Nitrogen Pesticides, ERA (Waters)
Standards Environmental Analysis Standards
One 2 mL flame-sealed ampule yields up to 2 L of this water pollution, nitrogen pesticides standard after dilution
Alachlor, Ametryn, Anilazine, Atraton, Atrazine, Bromacil, Butachlor, Butylate, Cyanazine, Deethyl atrazine, Deisopropyl atrazine, Diaminoatrazine, EPTC (Eptam), Hexazinone, Metolachlor, Metribuzin, Napropamide, Prometon, Prometryn, Pronamide, Propachlor, Propazine, Simazine, Terbacil, Trifluralin.
Use with EPA methods 619, 633, 8141 and 8270. This standard contains a subset of the analytes listed below at 2 to 20 µg/L.