Water Pollution Standard, Organochlorine Pesticides, ERA (Waters)
Supplier: ERA (Waters)
713
CA11022-790EA 660.22 CAD
CA11022-790
Water Pollution Standard, Organochlorine Pesticides, ERA (Waters)
Standards Environmental Analysis Standards
One 2 mL flame-sealed ampule yields up to 2 L of this water pollution, organochloride pesticides standard after dilution
Aldrin, alpha-BHC, beta-BHC, delta-BHC, gamma-BHC (Lindane), alpha-Chlordane, gamma-Chlordane, 4,4’-DDD, 4,4’-DDE, 4,4’-DDT, Dieldrin, Endosulfan I, Endosulfan II, Endosulfan sulfate, Endrin, Endrin aldehyde, Endrin ketone, Heptachlor, Heptachlor epoxide (beta), Methoxychlor.
Use with EPA methods 608 and 8081. This standard contains a subset of the analytes listed below at 1 to 20 µg/L.