Water Supply Standard, Metals, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
697 590
CA11020-084EA 381.14 CAD
CA11020-084 CA11022-740
Water Supply Standard, Metals, ERA (Waters)
Standards Environmental Analysis Standards
One 15 mL screw-cap vial yields up to 2 L of this water supply, metals standard after dilution
Use with ICP-OES, ICP-MS and AA methods.