Water Pollution Standard, PCBs in Water, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
734S 832S
CA11022-818EA 435.5 CAD
CA11022-818 CA11022-820
Water Pollution Standard, PCBs in Water, ERA (Waters)
Standards Environmental Analysis Standards
One 2 mL flame-sealed ampule yields up to 2 L of this water pollution, PCBs standard in water after dilution
Aroclor 1016, Aroclor 1221, Aroclor 1232, Aroclor 1242, Aroclor 1248, Aroclor 1254, Aroclor 1260.
Use with EPA methods 608 and 8082. This standard contains a different Aroclor, randomly selected from the list below at 2 to 10 µg/L.