Water Pollution Standard, Gasoline Range Organics (GRO) in Water, ERA (Waters)
Supplier: ENVIRONMENTAL RESOURCES ASS. (ERA)
762
CA11022-690EA 558.14 CAD
CA11022-690
Water Pollution Standard, Gasoline Range Organics (GRO) in Water, ERA (Waters)
Standards Environmental Analysis Standards
One 2 mL flame-sealed ampule yields up to 2 L of this water pollution, gasoline range organics (GRO) standard in water after dilution
Use with both purge and trap, as well as modified EPA 8015 GC/FID methods to test for GRO at 400 to 4,000 µg/L. This standard can also be used to test for BTEX in gasoline.